Cartridge, spe, Resprep WCX (Silica Based), (Ion-Exchange mode), High-quality, silica-based hydrophilic adsorbents, Used for common sample matrices such as food, soil, and water, WCX - weak cation exchange, Stable over the pH range (2 - 9), Size: 6mL/1000mg
Catalog #: 76470-254
Supplier: Restek
Cartridge, spe, Resprep WCX (Silica Based), (Ion-Exchange mode), High-quality, silica-based hydrophilic adsorbents, Used for common sample matrices such as food, soil, and water, WCX - weak cation exchange, Stable over the pH range (2 - 9), Size: 6mL/1000mg
Catalog #: 76470-254
Supplier: Restek
Customers Who Bought This Also Bought
n-Hexane ≥95.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$992.11 to $3,316.70
VWR® ARISTAR® Multi-Element ICP and ICP-MS Certified Reference Standards, Enhanced Packaging
Supplier: VWR International
$154.78
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
$31.63 to $756.61
Cryo Labels, Laser Printable, on Sheets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$176.71 to $184.67
SystemSURE Plus™ Luminometer, ATP Hygiene Monitoring System, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
$1,090.88
Copper(II) oxide, wire ACS (0.65x6 mm) for elementary analysis, Supelco®
Supplier: MilliporeSigma
$524.64 to $5,852.08