Cartridge, spe, Resprep Alumina N, High-quality, silica-based adsorbent, Used for common sample matrices such as food, soil, and petroleum, Certificates of analysis for this product are provided electronically, Size: 6mL/1000mg
Catalog #: 76470-242
Supplier: Restek
Customers Who Bought This Also Bought