Standard, d9-Tetrahydrocannabinolic Acid A (THCA-A), DEA-exempted formulation-no additional customer permits or licensing are required to purchase within the U.S, Certified reference materials (CRMs) manufactured and QC tested in ISO-accredited labs satisfy your ISO requirements, Size: 1 mL/ampul
Catalog #: 76470-226
Supplier: Restek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.