Quick-DNA HMW MagBead Kit, easiest method to purify high molecular weight DNA from any sample (including biological fluids, cells, solid tissue, environmental samples, High Molecular Weight DNA: Extract high molecular weight DNA up to 150 kb from sample, Highest DNA yield and purity, Size: 96 preps
Catalog #: 76469-432
Supplier: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.