ContactUsLinkComponent
Quick-RNA Viral Kit, quick, with zymospin, purification system for viral RNA from plasma, serum, cell culture media, cellular suspensions, urine, blood, saliva and any other biological samples stored in DNA/RNA Shield, Compatible with plasma, cell culture media, cellular suspensions, Size: 50 preps
Catalog #: 76469-372
Supplier:  Zymo Research
Quick-RNA Viral Kit, quick, with zymospin, purification system for viral RNA from plasma, serum, cell culture media, cellular suspensions, urine, blood, saliva and any other biological samples stored in DNA/RNA Shield, Compatible with plasma, cell culture media, cellular suspensions, Size: 50 preps
Catalog #: 76469-372
Supplier:  Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.