ContactUsLinkComponent
Filter Paper, Grade 740E, thick, absorbent cotton linter for use as an antibiotic assay test disc, Specified in standard tests for antibiotic susceptibility as well as antibiotic levels in milk, Coarse porosity, Fast flow rate, 20-30um particle retention, 12.7 mm circles
Catalog #: CA28446-006
Supplier:  Cytiva
Filter Paper, Grade 740E, thick, absorbent cotton linter for use as an antibiotic assay test disc, Specified in standard tests for antibiotic susceptibility as well as antibiotic levels in milk, Coarse porosity, Fast flow rate, 20-30um particle retention, 12.7 mm circles
Catalog #: CA28446-006
Supplier:  Cytiva
Customers Who Bought This Also Bought