Quick-RNA Viral 96 Kit, DX, Quick, high-throughput (96-well spin plate) purification of viral RNA from plasma, serum, CSF, cell culture media, cellular suspensions, urine, blood, saliva, swab, fecal, etc, provides for rapid, large scale isolation of viral RNA from plasma, serum, Size: 4x96 preps
Catalog #: 76469-092
Supplier: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.