Water Concentrating Buffer, Concentrates microbes, fungi and viral particles from water, Promotes stability of nucleic acids in water, All in one sample collection and concentration step, allows for the effective concentration of DNA and RNA from microbes, fungi, and viral particles, Size: 8ml
Catalog #: 76469-086
Supplier: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.