Environ Water RNA Kit, Enrich total RNA from large volumes (</-5 ml) and transform into small sample pellets for easy processing, simplifies COVID-19 wastewater surveillance by increasing viral enrichment, inactivating pathogens, and limits of detection, applications including RT-qPCR, Size: 50preps
Catalog #: 76469-084
Supplier: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.