ContactUsLinkComponent
Self-Contained Biological Indicator(SCBI) Ampoule, for monitoring steam sterilization processes, Spore carrier: Soybean casein digest growth medium with pH indicator, glass ampoule, Species: Geobacillus stearothermophilus, Mean population recovery: 1.0x10 to 6 - 5.0x10 to 6, pH indicator: Phenol red
Catalog #: 76470-192
Supplier:  Steris
Self-Contained Biological Indicator(SCBI) Ampoule, for monitoring steam sterilization processes, Spore carrier: Soybean casein digest growth medium with pH indicator, glass ampoule, Species: Geobacillus stearothermophilus, Mean population recovery: 1.0x10 to 6 - 5.0x10 to 6, pH indicator: Phenol red
Catalog #: 76470-192
Supplier:  Steris
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.