Spectrophotometer, Micro-Volume, with touchscreen and built-in sample ports for small volumes and cuvettes, quantifies very small amounts of DNA, RNA, oligos, and proteins, Increase sensitivity, Easy to use, built-in micro-volume sample port allows for working with sample volumes as small as 0.5 uL
Catalog #: MPCA7583898621
Supplier: MOLECULAR DEVICES LLC. CA
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.