ContactUsLinkComponent
Column, PD-10, empty disposable, Material: polypropylene, are recommended for use with bulk media, for affordable and versatile sample preparation, These are gravity operated sample preparation columns that cannot be connected to any system, Size: 15mm
Catalog #: MPCA29648002
Supplier:  Cytiva
Column, PD-10, empty disposable, Material: polypropylene, are recommended for use with bulk media, for affordable and versatile sample preparation, These are gravity operated sample preparation columns that cannot be connected to any system, Size: 15mm
Catalog #: MPCA29648002
Supplier:  Cytiva
Customers Who Bought This Also Bought