ContactUsLinkComponent
Vwr* Clear Starter kit, 3D Clear Reagent, Antibody, Blocking, Penetration, and Washing Buffers, Rapid tissue clearing, Easy to Use, No Special Equipment Required, Compatible with IF, FP and other Fluorescent lables, Microplate and Automation Compatible, Shelf Life: 24 months
Corning® 3D Clear Starter Kit, 3D Clear Reagent, Antibody, Blocking, Penetration and Washing Buffers, Corning
Catalog #: CA76463-340
Supplier:  VWR International
CAS Number:  
Vwr* Clear Starter kit, 3D Clear Reagent, Antibody, Blocking, Penetration, and Washing Buffers, Rapid tissue clearing, Easy to Use, No Special Equipment Required, Compatible with IF, FP and other Fluorescent lables, Microplate and Automation Compatible, Shelf Life: 24 months
Corning® 3D Clear Starter Kit, 3D Clear Reagent, Antibody, Blocking, Penetration and Washing Buffers, Corning
Catalog #: CA76463-340
Supplier:  VWR International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Corning® 3D Clear Starter Kit
  • Size:
    Kit
  • Cat. no.:
    CA76463-340
Specifications
About this item
Starter Kit combines Corning 3D Clear with buffer solutions required to process 3D tissue cultures.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent lables
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
The Starter Kit combines 30 ml of Corning 3D Clear with the four buffer solutions required to process 3D tissue cultures for immunolabeling.  All you need to do is add your immunolabels and nuclear stains. This kit comes with enough material to process three 96-well plates of 3D cell culture models.
Includes: Corning 3D Clear reagent (30 ml), Corning 3D Clear antibody buffer (30 ml), Corning 3D Clear blocking buffer (30 ml), Corning 3D Clear penetration buffer (30 ml), Corning 3D Clear washing buffer 10X (70 ml)
Caution: For best results, refrigerate buffer after opening.
Customers Who Bought This Also Bought