Order Entry
ContactUsLinkComponent
qScript 1-Step Virus ToughMix, Low ROX provides rapid detection of RNA viruses such as Flu-A, Flu-B, and SARS-COV-2. It has been optimized for maximum sensitivity to enable reliable quantification of very low input quantities, Size: 100 x 20 uL rxns (1 x 1 mL)
qScript™ 1-Step Virus ToughMix®, Virus Detection Kit, Quantabio
Catalog #: CA76463-380
Supplier:  Quantabio
CAS Number:  
qScript 1-Step Virus ToughMix, Low ROX provides rapid detection of RNA viruses such as Flu-A, Flu-B, and SARS-COV-2. It has been optimized for maximum sensitivity to enable reliable quantification of very low input quantities, Size: 100 x 20 uL rxns (1 x 1 mL)
qScript™ 1-Step Virus ToughMix®, Virus Detection Kit, Quantabio
Catalog #: CA76463-380
Supplier:  Quantabio
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
    RT-qPCR probe
  • Inhibitor-rich sample:
    Yes
  • Multiplexing:
    Yes
  • One step:
    Yes
  • Description:
    qScript™ 1-Step Virus ToughMix® low ROX
  • Cat. no.:
    CA76463-380
  • No. of Reactions:
    100

Specifications

About this item

Superior sensitivity for viral RNA detection.

  • Superior Sensitivity – Higher yields optimized for lower viral RNA inputs
  • Tough Tested - Tolerant to a wide range of PCR inhibitors
  • Multiplexing – Supports highly sensitive detection for up to four targets
  • Broad Dynamic Range – Reliable data from your precious samples every time

qScript™ 1-Step Virus ToughMix is a 2x, ready-to-use, master mix for rapid detection of RNA viruses such as Flu-A, Flu-B, and SARS-CoV-2, using one-step, or single-tube reverse transcription quantitative PCR (RT-qPCR). It has been optimized for maximum sensitivity to enable reliable quantification of very low input quantities of RNA using dual-labeled hydrolysis probe detection chemistries such as TaqMan® probes in single or multiplexed assay formats. qScript 1-Step Virus ToughMix contains all required components for RT-qPCR except RNA template and primer/probe assays.

qScript™ 1-Step Virus ToughMix is powered by an engineered reverse transcriptase with reduced RNase H activity and improved activity and stability at higher temperatures, that includes a RT Hot-Start technology to suppresses non-specific primer extension by the RT before cDNA synthesis. The use of higher temperatures (50 to 55 °C) for the first-strand step of one-step RT-qPCR provides higher specificity for primer annealing and disruption of RNA secondary structure that can interfere with cDNA synthesis. These features combined with a stringent mAb hot-start Taq polymerase and ToughMix reagent technology to neutralize many common PCR inhibitors, provide reproducible low-copy quantification as well as extended room temperature stability of fully assembled reactions for automated reaction assembly. qScript™ 1-Step Virus ToughMix does not contain an internal reference dye.

qScript™ 1-step Virus ToughMix was benchmarked against the gold standard qScript™ XLT 1-Step RT-qPCR ToughMix for detection of varying amounts of SARS-CoV-2 RNA in a background of human total RNA. Multiplexed RT-qPCR of the E-gene (FAM), RdRp (HEX) and RNase P Internal Control (ROX) was carried out on the Q qPCR cycler: 10 min at 50 °C; 1 min at 95 °C; followed by 45 cycles of 5 sec at 95 °C; 10 sec at 60 °C. Both products generated excellent results. However, the qScript 1-Step Virus ToughMix demonstrated earlier detection and robust amplification curves (higher signal amplitude) for low copy E-gene detection. Quantabio recommends both of these high performing products for RNA virus detection. While highly similar in composition, the improved specificity provided by the reverse transcription hot-start of the qScript™ 1-Step Virus ToughMix often translates to improved sensitivity and fewer false negative calls at or near the limit of detection (LOD), particularly in multiplexed assays.

Caution: qScript™ 1-Step Virus ToughMix® is intended for molecular biology applications. This product is not intended for the diagnosis, prevention or treatment of a disease.
Customers Who Bought This Also Bought