ContactUsLinkComponent
Autoclave bag, PreSterilized (gamma irradiated process), designed for Clean Room use. Printed BIOhazard bags meet all Federal, State, territorial regulations. Clear 2 mil (51UM) polypropylene for autoclave processing at 275 f (235 C). Packaged in 3 sealed sleeves of 50 bags with indicator, 31x38in
Catalog #: 76461-656
Supplier:  Tufpak
Autoclave bag, PreSterilized (gamma irradiated process), designed for Clean Room use. Printed BIOhazard bags meet all Federal, State, territorial regulations. Clear 2 mil (51UM) polypropylene for autoclave processing at 275 f (235 C). Packaged in 3 sealed sleeves of 50 bags with indicator, 31x38in
Catalog #: 76461-656
Supplier:  Tufpak
Customers Who Bought This Also Bought