ContactUsLinkComponent
Autoclave bag, PreSterilized (gamma irradiated process), designed for Clean Room applications. Clear 2 mil (51UM) polypropylene (plain clear bags) for processing at 275 deg f (235 deg C) in autoclave. Packaged in 2 sealed sleeves of 50 bags with GIP indicator, 37x48in
Catalog #: 76461-652
Supplier:  Tufpak
Autoclave bag, PreSterilized (gamma irradiated process), designed for Clean Room applications. Clear 2 mil (51UM) polypropylene (plain clear bags) for processing at 275 deg f (235 deg C) in autoclave. Packaged in 2 sealed sleeves of 50 bags with GIP indicator, 37x48in
Catalog #: 76461-652
Supplier:  Tufpak
Customers Who Bought This Also Bought