Discharge unit/Dispenser Head, Prospenser Plus, 3.0mm Spiral Tubing for FEP (Fluoroethylene propylene) compatible substances. Suitable for all Bottle-top Dispensers models.
Catalog #: CA76460-540
Supplier: Sartorius
Discharge unit/Dispenser Head, Prospenser Plus, 3.0mm Spiral Tubing for FEP (Fluoroethylene propylene) compatible substances. Suitable for all Bottle-top Dispensers models.
Catalog #: CA76460-540
Supplier: Sartorius
Customers Who Bought This Also Bought
Nalgene® Acrylic Burets, Automatic Self-Zeroing, PMP Stopcock, Thermo Scientific
Supplier: Thermo Fisher Scientific
$576.51 to $1,145.47
Nalgene® Self-Zeroing Buret Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
$177.89 to $307.53
Accessories for Costar® Transtar-96® Liquid Transfer System, Corning
Supplier: Corning
$462.01 to $462.01
Nalgene® Acrylic Burets, Class B, Polymethylpentene Stopcock, Thermo Scientific
Supplier: Thermo Fisher Scientific
$628.34 to $1,179.84
Axygen® Robotic Pipette Tips for AP96, AP384 and FX/NX Series Platforms, Corning
Supplier: Corning
$2,057.77 to $7,920.81
ART® Racks for Filter Tips Reload System, Thermo Scientific
Supplier: Thermo Scientific
$148.50 to $490.76