To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Calibration:Anycal 5 point customer buffer calibration
- Memory:4500 points
- Description:Digital handheld phenomenal® MD 8000 H multi-parameter meter, conductivity kit, with intelligent conductivity/temperature cell IDP 761-C with 1 bottle of 50 ml conductivity standard (1413 uS)
- Temperature accuracy:±0.2 °C
- Temperature resolution:0.1 °C
- Temperature range:auto: −5...+105 °C
- IP rating:IP 67
- Parameters:pH, mv, temperature, conductivity, dissolved oxygen, turbidity
- Calibration points:1 to 5 (automatic buffer recognition )
- Conductivity accuracy:±0.5% of value
- Conductivity cell constant:Automatic
- Conductivity range:0.01 µS/cm - 1000 mS/cm (depending on sensor)
- Connections:1 measurement input
- DO concentration:0.00 - 20.00 / 0.0 - 50.0 mg/L
- DO saturation:0.0 - 200.0%; 0 - 500%
- Data logging capacity:Up to 4500 data sets in automatic
- Data logging - log function:Manual/time controlled
- Display:Blacklit B/W graphic LCD display
- Interfaces:Mini USB-B (USB 1.1) for data downloads
- Reference temperature:20/25 °C
- Resistivity range:1 Ωcm - 20 MΩcm (depending on sensor)
- Salinity range:0 - 70
- TDS range:0 mg/L - 199.9 g/L
- mV accuracy:±0.3; ±1
- mV range:±2500; ±1200
- mV resolution:0.1; 1
- pH accuracy:±0.1, ±0.01, ±0.005
- pH range:0 - 14
- pH resolution:0.1/0.01/0.001
- Cat. no.:76460-512
Specifications
About this item
These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optimises use and makes measurement easier (no need to recalibrate when changing the meter, and no need to adjust the meter's settings). The measured data is complete with serial number, settings and calibration data.
- One universal measuring channel
- Digital sensor recognition: Measured parameter and settings are recognised automatically
- Suitable for Intelligent Digital Probes (IDP)
- Suitable for measurements with cable length up to 100 metres
- 4500-point memory
- Easy to read backlit B/W graphic display
- Low energy consumption (1000 operating hours)
76460-512 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml conductivity standard 0.01 mol/L KCl. Conductivity cell IDP 761-C and USB cable.
76460-514 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, optical DO sensor OPOX11-3 and USB cable.
76460-534 includes meter, case, stand, 4 LR06 batteries, 50 ml beaker, short instructions, CD ROM with manual and USB cable.