To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Bench top pH/mV/conductivity/DO meter pHenomenal® MU 6100L S1 kit
- Model:MU 6100L S1
- Temperature accuracy:±0.2
- Temperature resolution:0.1/1 °C
- Temperature range:Auto: −5.0...+10.0 °C; manual: −25...+130 °C
- Automatic buffer recognition:BDH Prolabo® TITRINORM® buffers at 25 °C: 4.00/6.98/9.94
NIST/DIN 19266 (1.68/4.00/6.86/9.18/12.54)
Technical buffers at 25 °C: 4.00/7.00/10.00 - Compliance:IP 43/GLP
- Conductivity accuracy:±0.5% of measured value
- Conductivity cell constant:0.001 - 25 cm⁻¹ (depending on probe)
- Conductivity range:0.001 µS/cm - 1000 mS/cm
- Conductivity resolution:0.1 µS/cm
- Connections:BNC for pH/banana for temperature/8-pin for conductivity and DO
- DO calibration:Saturated air
- DO concentration:0.00 - 20.00 / 0.0 - 90.0 mg/L
- DO concentration resolution:0.1%; 0.01 mg/L (depending on sensor)
- DO saturation:0.0 - 200.0%; 0 - 600% (depending on sensor)
- DO accuracy:±0.5% of value (between 5...30 °C)
- Data logging capacity:5000 data sets
- Data logging - log function:Selectable between 1 s and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
- Display:Graphic LCD with LED backlight
- Outputs:Waterproof USB connection (slave, bidirectional)
- Power supply:Mains supply or 4× AA batteries
- Resistivity accuracy:1 Ω/cm - 999 MΩ/cm (±0.5% of measured value)
- Salinity range:0.00 - 70 ppt
- TDS range:0 - 1999 mg/L / 2.00 - 19.99 g/L / 20.0 - 199.9 g/L
- TDS resolution:1 mg/L / 0.01 g/L / 0.1 g/L
- Temperature compensation:Automatic/manual
- mV accuracy:±0.3/±1
- mV range:±1200.0/±2500
- mV resolution:0.1/1
- pH accuracy:±0.1/±0.01/±0.005
- pH calibration:Up to 5 points
- pH range:−2.000...+19.999
- pH resolution:0.1/0.01/0.001 (selectable)
- Cat. no.:76460-494
Specifications
About this item
This user friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-parameter measurements. This meter can either measure pH/mV and conductivity or pH/mV and oxygen simultaneously. Complete with USB output for connection to a computer. GLP compliant.
- Large graphic LCD with continuous LED backlight shows the measured parameter and the temperature value simultaneously
- Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets
- Automatic power shut off: Adjustable from 10 min to 24 h
- Automatic buffer recognition of DIN and NIST buffers (1,68/4,00/6,86/9,18/12,54)
- Three additional technical buffers at 25 °C (4,00/7,00/10,00) in memory
- Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
- Low energy consumption. Choice of mains supply or 4× AA batteries for over 1000 operating hours
The MU 6100 L S2 kit (76460-496) includes meter, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, pH electrode pHenomenal 221, 50 ml conductivity standard 0.01 mol/L KCl, conductivity cell CO 11, 30 ml cleaning solution, 50 ml electrolyte, 3 exchange caps, DO electrode OXY 11 (3 m cable).
76460-520 includes meter, stand, 4 LR06 batteries, 50 ml beaker, short instructions, CD ROM with manual and plugin power supply.