ContactUsLinkComponent
Aqueous Cell, Material: Suprasil, Spectroscopic Quality, Flat Section Width: 16.9/70mm, When the sample contains water, the electric dipole moment of the water molecule interacts with the E field in the resonator, Dimension: 7x9mm, Lower Tube: 1x7-9mm, For: All TM 110 except JEOL, Volume: 275ul
Catalog #: 76460-236
Supplier:  SP Industries
Aqueous Cell, Material: Suprasil, Spectroscopic Quality, Flat Section Width: 16.9/70mm, When the sample contains water, the electric dipole moment of the water molecule interacts with the E field in the resonator, Dimension: 7x9mm, Lower Tube: 1x7-9mm, For: All TM 110 except JEOL, Volume: 275ul
Catalog #: 76460-236
Supplier:  SP Industries
Customers Who Bought This Also Bought