ContactUsLinkComponent
Aqueous Cell, Material: Suprasil, Spectroscopic Quality, Flat Section Width: 9.52mm, When the sample contains water, the electric dipole moment of the water molecule interacts with the E field in the resonator, Dimension: 5x7mm, Lower Tube: 1.5-2x7-9mm, For: All TE 102 except JEOL, Volume: 150ul
Catalog #: 76460-234
Supplier:  SP Industries
Aqueous Cell, Material: Suprasil, Spectroscopic Quality, Flat Section Width: 9.52mm, When the sample contains water, the electric dipole moment of the water molecule interacts with the E field in the resonator, Dimension: 5x7mm, Lower Tube: 1.5-2x7-9mm, For: All TE 102 except JEOL, Volume: 150ul
Catalog #: 76460-234
Supplier:  SP Industries
Customers Who Bought This Also Bought