Electrolytic Assembly, Material: Suprasil, complete cell includes two Calomel Reference Electrodes, a pair of Platinum Electrodes, and a gas bubbler for deoxygenation, For use with: JEOL, without Flat Cell Collar, Flat Section Width: 10mm, Flat Section Length: 60mm, Flat Section Sample Gap: 0.3mm
Catalog #: 76460-228
Supplier: SP Industries
Customers Who Bought This Also Bought