ContactUsLinkComponent
Aqueous Cell, Low Temperature, Material: Suprasil, Spectroscopic Quality, Small Volume TE102 Variable Temperature Aqueous Cell, Aqueous Cell works with most common VT Dewar Inserts, including WG-821(-X) and WG-848, For: All TE 102 or TE 011, Flat Section: 4.75/47mm, Upper Tube: 3x4mm, Volume: 100ul
Catalog #: 76460-226
Supplier:  SP Industries
Aqueous Cell, Low Temperature, Material: Suprasil, Spectroscopic Quality, Small Volume TE102 Variable Temperature Aqueous Cell, Aqueous Cell works with most common VT Dewar Inserts, including WG-821(-X) and WG-848, For: All TE 102 or TE 011, Flat Section: 4.75/47mm, Upper Tube: 3x4mm, Volume: 100ul
Catalog #: 76460-226
Supplier:  SP Industries
Customers Who Bought This Also Bought