Tissue Cell, Plate Material: Suprasil, used for viscous samples such as polymers and wet tissue, All cells, except WG-807-Q, include a 0.5 mm deep well for thick samples, sample is placed between the plates, Rod Length: 90/63mm, Cavity Dimension: 7x23mm, Baseplate: 16x65mm, Coverplate: 42x10mm
Catalog #: 76460-222
Supplier: SP Industries
Customers Who Bought This Also Bought