ContactUsLinkComponent
Vortex Plug, Finned, Thin Tube Wall Style, made of PTFE and chemcially resistent to most orgianic compound, Recommended for sample positioning inside the NMR tube, stainless steel positioning rod and recommend to use with WG-805, WG-805J, WG-805k, Dimension: 5x0.38 mm
Catalog #: 76460-218
Supplier:  SP Industries
Vortex Plug, Finned, Thin Tube Wall Style, made of PTFE and chemcially resistent to most orgianic compound, Recommended for sample positioning inside the NMR tube, stainless steel positioning rod and recommend to use with WG-805, WG-805J, WG-805k, Dimension: 5x0.38 mm
Catalog #: 76460-218
Supplier:  SP Industries
Customers Who Bought This Also Bought