Aqueous Flow Cell, Material: Suprasil, Continuous flow is a powerful tool to study free radical intermediates with short life-time at steady state. Wilmad-LabGlass manufactures two types of flow cells: room and variable temperature for such applications, sample thickness 0.25 mm in the flat section
Catalog #: 76460-216
Supplier: SP Industries
Customers Who Bought This Also Bought