ContactUsLinkComponent
Jet Tube Cleaner, Universal, with 24/40 Joint for 2.5 mm, can be used for any length NMR sample tube by a simple adjustment of the flexible PTFE tubing, By lifting the PTFE tubing out of the solvent reservoir, the sample tube can be air-dried, Fits Tubes with: 2.5-5mm
Catalog #: 76460-212
Supplier:  SP Industries
Jet Tube Cleaner, Universal, with 24/40 Joint for 2.5 mm, can be used for any length NMR sample tube by a simple adjustment of the flexible PTFE tubing, By lifting the PTFE tubing out of the solvent reservoir, the sample tube can be air-dried, Fits Tubes with: 2.5-5mm
Catalog #: 76460-212
Supplier:  SP Industries
Customers Who Bought This Also Bought