O-Ring, for Spinner, Uni-Finger, Red Viton, Causes less probe insert damage by better insert sample control, Variable temperature operation from -150 deg C to +200 deg C, Tachometer strip now with foil protection for improved life span, Weight compatible with room temperature spinners
Catalog #: 76460-016
Supplier: SP Industries
Customers Who Bought This Also Bought