ContactUsLinkComponent
NMR Tube, Thin Walled Precision, 350 MHZ, 10% more filling factor than economy tubes, For temperatures up to 230 degree C, and within a temperature step of 120 degree C, Ideal for experiments requiring critical shimming quality(high/ultrahigh field, non-spinning, multi-dimensional, Size: 5mmODx8in
Catalog #: 76459-856
Supplier:  SP Industries
NMR Tube, Thin Walled Precision, 350 MHZ, 10% more filling factor than economy tubes, For temperatures up to 230 degree C, and within a temperature step of 120 degree C, Ideal for experiments requiring critical shimming quality(high/ultrahigh field, non-spinning, multi-dimensional, Size: 5mmODx8in
Catalog #: 76459-856
Supplier:  SP Industries
Customers Who Bought This Also Bought