SP Wilmad-LabGlass NMR Tube Caps, PTFE, SP Industries
Catalog #: 76459-690
Supplier: SP Industries
Specifications
- Color:White
- Material:PTFE
- Cat. no.:76459-690
- For Use With:Wilmad Precision NMR tubes
- For Tube O.D.:5 mm
Specifications
About this item
The PTFE NMR tube caps are machined to exact specifications with a smaller gyroradius and more homogeneous mass distribution than disposable caps for better spinning stability.
These caps are recommended in experiments at high to ultra high field and experiments using chloroform as reference solution.
Caution: Only to be used with Wilmad precision NMR tubes.
Customers Who Bought This Also Bought