SP Wilmad-LabGlass Disposable NMR Tube Caps, SP Industries
Catalog #: 76459-652
Supplier: SP Industries
Specifications
- Color:Blue
- Material:Polyethylene
- Cat. no.:76459-652
- For Tube O.D.:10 mm
Specifications
About this item
Manufactured from high quality polyethylene or ethylene vinyl acetate to ensure a tight seal.
- Different colors help to track samples
- Available in large quantities for high throughput laboratories
Caution: Please avoid using disposable NMR tube caps when CDCl₃ serves as the reference solution as the material(s) could be dissolved. For CDCl₃, we recommend PTFE tube caps.
Customers Who Bought This Also Bought