ContactUsLinkComponent
Coaxial System Complete, with 2.97mm Insert Tube, Switch between three unique sample ratios during external referencing experiments, temperature experiments as materials is the same between the tubes and spacer, Complete set come with inner and outer tubes, spacer and cap with hole, Size: 4.97 mm OD
Catalog #: 76459-640
Supplier:  SP Industries
Coaxial System Complete, with 2.97mm Insert Tube, Switch between three unique sample ratios during external referencing experiments, temperature experiments as materials is the same between the tubes and spacer, Complete set come with inner and outer tubes, spacer and cap with hole, Size: 4.97 mm OD
Catalog #: 76459-640
Supplier:  SP Industries
Customers Who Bought This Also Bought