ContactUsLinkComponent
Coaxial system, with 3.30 mm insert tube, Switch between three unique sample ratios during external referencing experiment, Ideal for variable temperature experiments as materials is the same between the tubes and spacer, Complete sets come with outer tubes, spacer and cap with hole, Size: 4.97mm OD
Catalog #: 76459-638
Supplier:  SP Industries
Coaxial system, with 3.30 mm insert tube, Switch between three unique sample ratios during external referencing experiment, Ideal for variable temperature experiments as materials is the same between the tubes and spacer, Complete sets come with outer tubes, spacer and cap with hole, Size: 4.97mm OD
Catalog #: 76459-638
Supplier:  SP Industries
Customers Who Bought This Also Bought