ContactUsLinkComponent
Coaxial System Complete, 600MHz, provides extra protection for toxic sample. It can also be used with a reference standard which is insoluable in the sample or may cause a reaction. There is approximately a 50 um gap between the inner and outer tubes, Ideal for variable temperature, Size: 5 mm
Catalog #: 76459-636
Supplier:  SP Industries
Coaxial System Complete, 600MHz, provides extra protection for toxic sample. It can also be used with a reference standard which is insoluable in the sample or may cause a reaction. There is approximately a 50 um gap between the inner and outer tubes, Ideal for variable temperature, Size: 5 mm
Catalog #: 76459-636
Supplier:  SP Industries
Customers Who Bought This Also Bought