Exam Gloves, 93-732, 1.5 AQL, Material: Nitrile, Color: Black, Chlorinated, Silicone Free, Textured Fingers, Beaded Cuff, Enhanced dexterity and tactility in a thin, yet strong design, Silicone free to ensure advanced product protection, Length: 9.6inch, Size: XL (9.5 - 10)
Catalog #: 76459-138
Supplier: Ansell Healthcare
Customers Who Bought This Also Bought