ContactUsLinkComponent
VWR Vacuum pump connection kit - for vacuum pump VAP1 and VAP2; consisting of aluminum clamping ring, universal centering ring, small flange with hose nozzle, (2.5 m) rubber hose and 2 hose clamps, for: VD 23, VD 56, VD 115 Vacuum drying chambers
Catalog #: 76458-976
Supplier:  Binder
VWR Vacuum pump connection kit - for vacuum pump VAP1 and VAP2; consisting of aluminum clamping ring, universal centering ring, small flange with hose nozzle, (2.5 m) rubber hose and 2 hose clamps, for: VD 23, VD 56, VD 115 Vacuum drying chambers
Catalog #: 76458-976
Supplier:  Binder
Customers Who Bought This Also Bought