ContactUsLinkComponent
NMR Tube, Thin Wall Precision, PTFE Coated, has excellent mechanical strength that can survive a 6 foot free drop at any angle, PTFE coating also yields zero background in proton spectrum, these two features make it ideal in food and pharmaceutical studies, Dimension: 5 mm, Length: 7inch
Catalog #: 76460-142
Supplier:  SP Industries
NMR Tube, Thin Wall Precision, PTFE Coated, has excellent mechanical strength that can survive a 6 foot free drop at any angle, PTFE coating also yields zero background in proton spectrum, these two features make it ideal in food and pharmaceutical studies, Dimension: 5 mm, Length: 7inch
Catalog #: 76460-142
Supplier:  SP Industries
Customers Who Bought This Also Bought