ContactUsLinkComponent
Tube Washer, NMR, After fitting the washer to a filter flask, an inverted sample tube is inserted into the washer and solvent is introduced into the reservoir using a series of wash bottles, single tube washers require flasks with vacuum side arms, Dimension: 5x10mm, Tube Length: 8inch
Catalog #: 76460-130
Supplier:  SP Industries
Tube Washer, NMR, After fitting the washer to a filter flask, an inverted sample tube is inserted into the washer and solvent is introduced into the reservoir using a series of wash bottles, single tube washers require flasks with vacuum side arms, Dimension: 5x10mm, Tube Length: 8inch
Catalog #: 76460-130
Supplier:  SP Industries
Customers Who Bought This Also Bought