Mass Multiplier Ring, Material: PEEK, with 1 or 2 O-Rings at the top, Cost effective compared to the full VT version for sample changers, Includes high temperature external O-rings at the top and bottom for long life and a firm grip on the sample at top and bottom, for V-NS Style Spinner,
Catalog #: 76460-120
Supplier: SP Industries
Customers Who Bought This Also Bought