O-Ring, For Spinner STV-5 & V-PEEK-5, Color: Red, Causes less probe insert damage by better insert sample control, Tachometer strip now with foil protection for improved life span, Longer upper barrel stabilizer easily identified by 3mm yellow band, Susceptibility matched to the originals
Catalog #: 76460-118
Supplier: SP Industries
Customers Who Bought This Also Bought