cryo rack, Material: Polypropylene, Colour: black, high rack with lid, code 128, s1, centered, human readable number, datamatrix code on rack bottom, for 48 cryo tubes 1272xx, non-sterile, Size: 4ml
Catalog #: 76458-330
Supplier: Greiner Bio-One
Customers Who Bought This Also Bought