ContactUsLinkComponent
Tube set, LPV Shigemi, CD3OD, 12mm Bottom, Standard full sets include one outer tube and one inner insert. Gas-Tight sets include one outer tube fused with a complete LPV valve, one inner insert with detachable PTFE Holder and one Positioning rod, Dimension: 5MM
Catalog #: 76458-258
Supplier:  SP Industries
Tube set, LPV Shigemi, CD3OD, 12mm Bottom, Standard full sets include one outer tube and one inner insert. Gas-Tight sets include one outer tube fused with a complete LPV valve, one inner insert with detachable PTFE Holder and one Positioning rod, Dimension: 5MM
Catalog #: 76458-258
Supplier:  SP Industries
Customers Who Bought This Also Bought