ContactUsLinkComponent
Tube set, Shigemi, CD3OD, 15mm Bottom, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 10MM
Catalog #: 76458-230
Supplier:  SP Industries
Tube set, Shigemi, CD3OD, 15mm Bottom, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 10MM
Catalog #: 76458-230
Supplier:  SP Industries
Customers Who Bought This Also Bought