ContactUsLinkComponent
Tube set, Shigemi, CD3OD Universal, 10mm Bottom, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 3MM
Catalog #: 76458-218
Supplier:  SP Industries
Tube set, Shigemi, CD3OD Universal, 10mm Bottom, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 3MM
Catalog #: 76458-218
Supplier:  SP Industries
Customers Who Bought This Also Bought