ContactUsLinkComponent
Tube set, Shigemi, DMSO-D6, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 8MM
Catalog #: 76458-204
Supplier:  SP Industries
Tube set, Shigemi, DMSO-D6, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 8MM
Catalog #: 76458-204
Supplier:  SP Industries
Customers Who Bought This Also Bought