Tube set, Shigemi, THIN D20, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 5MM
Catalog #: 76458-170
Supplier: SP Industries
Customers Who Bought This Also Bought