ContactUsLinkComponent
Tube set, Shigemi, Deut OX D2O, 15mm Bottom, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 5MM
Catalog #: 76458-158
Supplier:  SP Industries
Tube set, Shigemi, Deut OX D2O, 15mm Bottom, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 5MM
Catalog #: 76458-158
Supplier:  SP Industries
Customers Who Bought This Also Bought