ContactUsLinkComponent
Tube set, Shigemi, Deut OX D2O, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 4MM
Catalog #: 76458-148
Supplier:  SP Industries
Tube set, Shigemi, Deut OX D2O, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length, Dimension: 4MM
Catalog #: 76458-148
Supplier:  SP Industries
Customers Who Bought This Also Bought