ContactUsLinkComponent
Tube set, matched with Acetone-d6, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length,Dimension: 2.5MM
Catalog #: 76458-128
Supplier:  SP Industries
Tube set, matched with Acetone-d6, feature would effectively reduce the boundary gradients at the edges of the coil, therefore, it is no longer critical to keep the general NMR sample rule as sample height = 3 times the coil length,Dimension: 2.5MM
Catalog #: 76458-128
Supplier:  SP Industries
Customers Who Bought This Also Bought