MS Nut, for Shimadzu 2030 GC-MS instruments with QP2020 NX or QP2010 SE mass spectrometers, 1.2 mm nut bore permits easy removal of ferrules with a standard tapered-needle file, Made of brass to prevent thread stripping on the transfer line, Design enhances ease of threading onto the transfer line
Catalog #: 76458-102
Supplier: Restek
Customers Who Bought This Also Bought